Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190852.1 | NP_001177781.1 | 3 | 338 | |
NM_001190853.1 | NP_001177782.1 | 3 | 338 | ||
NM_001190854.1 | NP_001177783.1 | 3 | 338 | ||
NM_001190855.1 | NP_001177784.1 | 3 | 338 | ||
NM_001190856.1 | NP_001177785.1 | 3 | 338 | ||
NM_001190857.1 | NP_001177786.1 | 2 | 227 | ||
NM_019808.3 | NP_062782.2 | 3 | 338 | ||
NM_019809.3 | NP_062783.2 | 3 | 338 | ||
NM_022554.3 | NP_072048.2 | 2 | 227 | ||
GenBank mRNA | AA061806.1 | - | 1 | 24 | |
AB016586.1 | - | 3 | 260 | ||
AB016587.1 | - | 2 | 239 | ||
AB016588.1 | - | 3 | 260 | ||
AK003194.1 | - | 3 | 281 | ||
AK009464.1 | - | 3 | 250 | ||
AK089779.1 | - | 3 | 338 | ||
AK147999.1 | - | 3 | 278 | ||
BC037476.1 | - | 3 | 302 | 4457868 | |
BY227455.1 | - | 3 | 309 | ||
BY741234.1 | - | 3 | 274 | ||
CA534475.1 | - | 2 | 200 | ||
CB597177.1 | - | 3 | 269 | ||
DQ177283.1 | - | 2 | 185 | ||
HM369794.1 | - | 2 | 185 | ||
HM369795.1 | - | 2 | 185 | ||
HM369796.1 | - | 2 | 185 | ||
HM369797.1 | - | 2 | 185 | ||
HM369798.1 | - | 2 | 185 |