Gene Transcripts

Gene Symbol
Vmn2r26
Entrez Gene ID
56552
Gene Name
vomeronasal 2, receptor 26
Gene Aliases
V2R1, V2r1b
Chromosome Location
Chr. 6: 124024758 - 124062035 on Build GRCm38
UniGene ID
Mm.23795
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019917.2 NP_064301.2 2 462
GenBank mRNA AF053985.1 - 2 462
AY426342.1 - 2 462

Gene Symbol
Vmn2r24
Entrez Gene ID
243628
Gene Name
vomeronasal 2, receptor 24
Gene Aliases
EG243628
Chromosome Location
Chr. 6: 123778971 - 123816280 on Build GRCm38
UniGene ID
Mm.469873
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104639.1 NP_001098109.1 2 462

Gene Symbol
Vmn2r25
Entrez Gene ID
545874
Gene Name
vomeronasal 2, receptor 25
Gene Aliases
EG545874
Chromosome Location
Chr. 6: 123822814 - 123853190 on Build GRCm38
UniGene ID
Mm.347595
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104641.1 NP_001098111.1 2 462

View Full Product Details