Gene Transcripts

Gene Symbol
Slc16a8
Entrez Gene ID
57274
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 8
Gene Aliases
Mct3
Chromosome Location
Chr. 15: 79251016 - 79254748 on Build GRCm38
UniGene ID
Mm.441697
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020516.2 NP_065262.1 4 500
GenBank mRNA AF019111.2 - 4 509
BC018216.1 - 4 500 4481698

View Full Product Details