Gene Transcripts

Gene Symbol
Chml
Entrez Gene ID
12663
Gene Name
choroideremia-like
Gene Aliases
AI450814, E030003F13Rik, REP-2, Rep2
Chromosome Location
Chr. 1: 175682237 - 175688353 on Build GRCm38
UniGene ID
Mm.440490
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021350.2 NP_067325.2 1 127
GenBank mRNA BC118932.2 - 1 127

View Full Product Details