Gene Transcripts
- Gene Symbol
- Rangrf
- Entrez Gene ID
- 57785
- Gene Name
- RAN guanine nucleotide release factor
- Gene Aliases
- 2400006H24Rik, Mog1, Rangnrf
- Chromosome Location
- Chr. 11: 68972484 - 68975185 on Build GRCm38
- UniGene ID
- Mm.143774
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285441.1 |
NP_001272370.1
|
3, 4 |
400 |
|
NM_001285442.1 |
NP_001272371.1
|
3, 4 |
475 |
|
NM_021329.3 |
NP_067304.1
|
3, 4 |
496 |
|
GenBank mRNA |
AF243512.1 |
- |
3, 4 |
467 |
|
AK010288.1 |
- |
3, 4 |
496 |
|
BB618250.1 |
- |
3, 4 |
494 |
|
BC018290.1 |
- |
3, 4 |
399 |
|
BC038470.1 |
- |
3, 4 |
477 |
5344659 |
BC087921.1 |
- |
3, 4 |
461 |
6772909 |
CF751505.1 |
- |
3, 4 |
328 |
|
View Full Product Details