Gene Transcripts
- Gene Symbol
- Nkain4
- Entrez Gene ID
- 58237
- Gene Name
- Na+/K+ transporting ATPase interacting 4
- Gene Aliases
- AB030182, C030019F02Rik
- Chromosome Location
- Chr. 2: 180934769 - 180954888 on Build GRCm38
- UniGene ID
- Mm.40361
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001141933.1 |
NP_001135405.1
|
3, 4 |
356 |
|
NM_021426.4 |
NP_067401.3
|
3, 4 |
356 |
|
GenBank mRNA |
AB030182.1 |
- |
3, 4 |
334 |
|
AK005262.1 |
- |
3, 4 |
327 |
|
AK007632.1 |
- |
3, 4 |
321 |
|
AK021096.1 |
- |
3, 4 |
351 |
|
BC061107.1 |
- |
3, 4 |
341 |
30295742 |
BC089330.1 |
- |
3, 4 |
308 |
30358178 |
CK625327.1 |
- |
3, 4 |
356 |
|
EF058055.1 |
- |
3, 4 |
281 |
|
EF058056.1 |
- |
3, 4 |
281 |
|
EF058057.1 |
- |
3, 4 |
281 |
|
View Full Product Details