Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164197.1 | NP_001157669.1 | 2 | 108 | |
NM_021412.2 | NP_067387.1 | 2 | 243 | ||
GenBank mRNA | AF153199.1 | - | 2 | 195 | |
AF155221.1 | - | 2 | 223 | ||
AF162446.1 | - | 2 | 223 | ||
AK079924.1 | - | 2 | 109 | ||
AK080125.1 | - | 2 | 103 | ||
AK136517.1 | - | 2 | 97 | ||
AK154863.1 | - | 2 | 236 | ||
BC112966.2 | - | 2 | 215 | 40039023 |