Gene Transcripts

Gene Symbol
Tmem8
Entrez Gene ID
60455
Gene Name
transmembrane protein 8 (five membrane-spanning domains)
Gene Aliases
AW490096, M83, Tmem8a
Chromosome Location
Chr. 17: 26112691 - 26123254 on Build GRCm38
UniGene ID
Mm.304656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021793.2 NP_068565.1 3 525
GenBank mRNA AB045293.1 - 3 524
AK044384.1 - 3 526
BC005722.1 - 2 401 3595205

View Full Product Details