Gene Transcripts

Gene Symbol
Chst12
Entrez Gene ID
59031
Gene Name
carbohydrate sulfotransferase 12
Gene Aliases
AI595374, C4S-2, C4ST2
Chromosome Location
Chr. 5: 140505580 - 140525238 on Build GRCm38
UniGene ID
Mm.28934
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021528.3 NP_067503.3 2 886
GenBank mRNA AJ289132.1 - 1 799
AK142155.1 - 2 884
AK163588.1 - 2 856
AK170209.1 - 2 849
AK170484.1 - 2 853
AK171208.1 - 2 853
BC003201.1 - 2 867 3586766

View Full Product Details