Gene Transcripts

Gene Symbol
Cenpk
Entrez Gene ID
60411
Gene Name
centromere protein K
Gene Aliases
B130045K24Rik, BB232189, C530004N04Rik, Cenp-K, Solt, Solzt
Chromosome Location
Chr. 13: 104228611 - 104249622 on Build GRCm38
UniGene ID
Mm.281498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021790.2 NP_068562.1 8, 9 913
NM_181061.5 NP_851406.1 8, 9 1036
GenBank mRNA AB043687.1 - 8, 9 913
AK082939.1 - 8, 9 693
AK135773.1 - 429
BC028655.1 - 8, 9 1049

View Full Product Details