Gene Transcripts

Gene Symbol
Abhd8
Entrez Gene ID
64296
Gene Name
abhydrolase domain containing 8
Gene Aliases
0910001L24Rik, AB030191
Chromosome Location
Chr. 8: 71456700 - 71463657 on Build GRCm38
UniGene ID
Mm.276383
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022419.3 NP_071864.2 2 862
GenBank mRNA AB030191.1 - 2 852
AK003090.1 - 2 850
AK037669.1 - 2 828
AK080618.1 - 2 861
AK155503.1 - 2 862
AK181179.1 - 1 49
AK205860.1 - 1 170
BC026540.1 - 2 811 5050705

View Full Product Details