Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023418.2 | NP_075907.2 | 2 | 518 | |
GenBank mRNA | AF283667.1 | - | 2 | 430 | |
AK004921.1 | - | 2 | 374 | ||
AK009905.2 | - | 2 | 371 | ||
AK144681.1 | - | 2 | 372 | ||
AK150716.1 | - | 2 | 367 | ||
AK152443.1 | - | 2 | 367 | ||
AK153202.1 | - | 2 | 371 | ||
AK159305.1 | - | 2 | 371 | ||
AK165410.1 | - | 2 | 367 | ||
AK168949.1 | - | 2 | 364 | ||
AK192398.1 | - | 1 | 149 | ||
AK211756.1 | - | 1 | 63 | ||
AK212069.1 | - | 1 | 63 | ||
AK216443.1 | - | 1 | 63 | ||
BC002241.1 | - | 2 | 466 | 3491964 | |
BC005661.1 | - | 2 | 432 | 3588761 | |
BC066844.1 | - | 2 | 355 | 30471755 | |
BC067412.1 | - | 1 | 171 | ||
BC083090.1 | - | 2 | 367 | 30444229 | |
BC106139.1 | - | 2 | 438 | 6528521 |