Gene Transcripts

Gene Symbol
Wdr31
Entrez Gene ID
71354
Gene Name
WD repeat domain 31
Gene Aliases
5430402I10Rik, spWD
Chromosome Location
Chr. 4: 62448653 - 62470964 on Build GRCm38
UniGene ID
Mm.155573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290521.1 NP_001277450.1 3, 4 435
NM_001290522.1 NP_001277451.1 3, 4 325
NM_023597.2 NP_076086.1 3, 4 328
GenBank mRNA AF274321.1 - 3, 4 305
AK017259.1 - 3, 4 318
AK141026.1 - 3, 4 433
BC026500.1 - 2, 3 214
BC037102.1 - 3, 4 318 5364307
CK618879.1 - 3, 4 435

View Full Product Details