Gene Transcripts
- Gene Symbol
- Ost4
- Entrez Gene ID
- 67695
- Gene Name
- oligosaccharyltransferase 4 homolog (S. cerevisiae)
- Gene Aliases
- 2310016E02Rik
- Chromosome Location
- Chr. 5: 30906516 - 30907807 on Build GRCm38
- UniGene ID
- Mm.21450
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134692.2 |
NP_001128164.1
|
1, 2 |
398 |
|
NM_024460.4 |
NP_077780.3
|
2, 3 |
270 |
|
GenBank mRNA |
AA563474.1 |
- |
2, 3 |
270 |
|
AK002224.2 |
- |
2, 3 |
245 |
|
AK002453.1 |
- |
2, 3 |
192 |
|
AK003569.1 |
- |
2, 3 |
249 |
|
AK005413.1 |
- |
2, 3 |
193 |
|
AK009384.1 |
- |
|
205 |
|
AK010297.1 |
- |
1, 2 |
334 |
|
AK019285.1 |
- |
2, 3 |
230 |
|
BC002177.1 |
- |
1, 2 |
335 |
3486998 |
BC082316.1 |
- |
1, 2 |
309 |
6814413 |
BY089912.1 |
- |
1, 2 |
332 |
|
CA466367.1 |
- |
2, 3 |
228 |
|
DV041491.1 |
- |
|
311 |
|
View Full Product Details