Gene Transcripts
- Gene Symbol
- ARSE
- Entrez Gene ID
- 415
- Gene Name
- arylsulfatase E (chondrodysplasia punctata 1)
- Gene Aliases
- ASE, CDPX, CDPX1, CDPXR
- Chromosome Location
- Chr. X: 2934632 - 2968310 on Build GRCh38
- UniGene ID
- Hs.386975
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000047.2 |
NP_000038.2
|
3, 4 |
443 |
|
NM_001282628.1 |
NP_001269557.1
|
4, 5 |
729 |
|
XM_005274518.2 |
XP_005274575.1
|
3, 4 |
370 |
|
XM_005274519.4 |
XP_005274576.1
|
3, 4 |
533 |
|
XM_011545521.1 |
XP_011543823.1
|
3, 4 |
574 |
|
XM_017029525.1 |
XP_016885014.1
|
4, 5 |
660 |
|
XM_017029526.1 |
XP_016885015.1
|
4, 5 |
774 |
|
GenBank mRNA |
AK223183.1 |
- |
3, 4 |
273 |
|
AK223199.1 |
- |
3, 4 |
240 |
|
AK301004.1 |
- |
4, 5 |
560 |
|
AK313093.1 |
- |
3, 4 |
262 |
|
BC130438.1 |
- |
3, 4 |
238 |
40146469 |
BP272004.1 |
- |
3, 4 |
375 |
|
BP272164.1 |
- |
3, 4 |
376 |
|
DA915010.1 |
- |
3, 4 |
445 |
|
DB073314.1 |
- |
3, 4 |
425 |
|
X83573.1 |
- |
3, 4 |
260 |
|
View Full Product Details