Gene Transcripts

Gene Symbol
PRKCQ
Entrez Gene ID
5588
Gene Name
protein kinase C theta
Gene Aliases
PRKCT, nPKC-theta
Chromosome Location
Chr. 10: 6393038 - 6580646 on Build GRCh38
UniGene ID
Hs.498570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242413.2 NP_001229342.1 3 306
NM_001282644.1 NP_001269573.1 3 304
NM_001282645.1 NP_001269574.1 3 306
NM_001323265.1 NP_001310194.1 3 361
NM_001323266.1 NP_001310195.1 3 304
NM_001323267.1 NP_001310196.1 3 304
NM_006257.4 NP_006248.1 3 306
XM_005252496.4 XP_005252553.1 3 406
XM_005252497.4 XP_005252554.1 3 408
XM_006717465.3 XP_006717528.1 5 2801
XM_017016410.1 XP_016871899.1 4 2712
XM_017016411.1 XP_016871900.1 5 2801
GenBank mRNA AK293935.1 - 3 244
AK308099.1 - 3 279
AK312411.1 - 3 256
AU099414.1 - 3 281
BC101465.1 - 2 225 8068971
BC113359.1 - 2 225 8322411
BX647657.1 - 2 390
L01087.1 - 3 300
L07032.1 - 2 227

View Full Product Details