Gene Transcripts

Gene Symbol
Gid4
Entrez Gene ID
66771
Gene Name
GID complex subunit 4, VID24 homolog (S. cerevisiae)
Gene Aliases
4933439F18Rik
Chromosome Location
Chr. 11: 60417145 - 60445277 on Build GRCm38
UniGene ID
Mm.29950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025757.4 NP_080033.3 2, 3 757
GenBank mRNA AJ404329.1 - 2, 3 245
AK012042.1 - 2, 3 585
AK017120.1 - 757
AK039435.1 - 2, 3 641
AK077759.1 - 2, 3 324
AK142753.1 - 2, 3 559
AK164644.1 - 2, 3 624
AK196423.1 - 1, 2 55
BC044901.1 - 2, 3 459 3660276
BC046821.1 - 2, 3 571 6400140
BC052418.1 - 2, 3 599 6401069

View Full Product Details