Gene Transcripts

Gene Symbol
Gid4
Entrez Gene ID
66771
Gene Name
GID complex subunit 4, VID24 homolog (S. cerevisiae)
Gene Aliases
4933439F18Rik
Chromosome Location
Chr. 11: 60417145 - 60445277 on Build GRCm38
UniGene ID
Mm.29950
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025757.4 NP_080033.3 2, 3 768
GenBank mRNA AJ404329.1 - 2, 3 256
AK012042.1 - 2, 3 596
AK017120.1 - 768
AK039435.1 - 2, 3 652
AK077759.1 - 2, 3 335
AK142753.1 - 2, 3 570
AK164644.1 - 2, 3 635
AK196423.1 - 1, 2 66
BC044901.1 - 2, 3 470 3660276
BC046821.1 - 2, 3 582 6400140
BC052418.1 - 2, 3 610 6401069

View Full Product Details