Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026034.4 | NP_080310.1 | 4 | 618 | |
GenBank mRNA | AK009821.1 | - | 4 | 612 | |
AK011303.1 | - | 4 | 617 | ||
AK012872.1 | - | 4 | 593 | ||
AK015272.1 | - | 4 | 751 | ||
BC027343.1 | - | 4 | 549 | ||
BC038487.1 | - | 4 | 570 | 2812558 | |
BC046281.1 | - | 4 | 570 | 6395384 | |
BC058573.1 | - | 4 | 659 | 6487440 |