Gene Transcripts

Gene Symbol
Gatad1
Entrez Gene ID
67210
Gene Name
GATA zinc finger domain containing 1
Gene Aliases
2310031E19Rik, 2810047M21Rik, 8430439A17Rik, 9130430G15Rik, B330017N08Rik, ENSMUSG00000073241, Odag
Chromosome Location
Chr. 5: 3639961 - 3647936 on Build GRCm38
UniGene ID
Mm.178424
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026033.2 NP_080309.2 5 1433
GenBank mRNA AB047921.1 - 5 1414
AK012921.1 - 5 1434
AK078416.1 - 5 1378

View Full Product Details