Gene Transcripts

Gene Symbol
Unc50
Entrez Gene ID
67387
Gene Name
unc-50 homolog (C. elegans)
Gene Aliases
1110002A21Rik, GMH1, UNCL, URP
Chromosome Location
Chr. 1: 37430144 - 37439124 on Build GRCm38
UniGene ID
Mm.27404
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026123.3 NP_080399.1 2 444
GenBank mRNA AK003273.1 - 2 219
AK004433.1 - 2 208
AK132757.1 - 2 445
AK135885.1 - 2 436
AK217920.1 - 2 166
AY017214.1 - 2 429
BC019484.1 - 2 411 4039903

View Full Product Details