Gene Transcripts

Gene Symbol
Ssr3
Entrez Gene ID
67437
Gene Name
signal sequence receptor, gamma
Gene Aliases
0610038P07Rik, AL022999, AU022074, AW553833, TRAPG
Chromosome Location
Chr. 3: 65379657 - 65392553 on Build GRCm38
UniGene ID
Mm.232728
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026155.3 NP_080431.1 3 303
GenBank mRNA AK002818.1 - 3 304
AK029353.1 - 3 304
AK076235.1 - 3 336
AK088555.1 - 3 308
AK150255.1 - 3 297
AK192841.1 - 3 224
AK194871.1 - 3 224
AK199053.1 - 3 224
AK206034.1 - 3 224
BC011111.1 - 3 310 3965897
BC020335.1 - 3 275
CT010172.1 - 3 287

View Full Product Details