Gene Transcripts

Gene Symbol
Gpatch1
Entrez Gene ID
67471
Gene Name
G patch domain containing 1
Gene Aliases
1300003A17Rik, ECGP, Gpatc1
Chromosome Location
Chr. 7: 35276544 - 35318440 on Build GRCm38
UniGene ID
Mm.358705
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026181.1 NP_080457.1 20 3029
GenBank mRNA AK004868.1 - 20 3029
AK154571.1 - 21 2962
BC026394.1 - 14 2195
BC100577.1 - 20 3004

View Full Product Details