Gene Transcripts
- Gene Symbol
- FUCA1
- Entrez Gene ID
- 2517
- Gene Name
- fucosidase, alpha-L- 1, tissue
- Gene Aliases
- FUCA
- Chromosome Location
- Chr. 1: 23845077 - 23868369 on Build GRCh38
- UniGene ID
- Hs.370858
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000147.4 |
NP_000138.2
|
4, 5 |
859 |
|
XM_005245821.2 |
XP_005245878.1
|
4, 5 |
996 |
|
XM_011541167.2 |
XP_011539469.1
|
4, 5 |
444 |
|
XM_017000905.1 |
XP_016856394.1
|
4, 5 |
541 |
|
GenBank mRNA |
AB074175.1 |
- |
4, 5 |
780 |
|
AK095443.1 |
- |
5, 6 |
731 |
|
AK095455.1 |
- |
5, 6 |
745 |
|
AK225468.1 |
- |
4, 5 |
780 |
|
AK301302.1 |
- |
6, 7 |
709 |
|
AK310143.1 |
- |
5, 6 |
842 |
|
AK314649.1 |
- |
4, 5 |
780 |
|
BC017338.2 |
- |
4, 5 |
782 |
4871788 |
M10355.1 |
- |
4, 5 |
554 |
|
M29877.1 |
- |
4, 5 |
779 |
|
X01390.1 |
- |
4, 5 |
554 |
|
View Full Product Details