Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163316.1 | NP_001156788.1 | 5, 6 | 552 | |
NM_026269.2 | NP_080545.2 | 5, 6 | 743 | ||
GenBank mRNA | AK003551.1 | - | 5, 6 | 692 | |
BC019557.1 | - | 5, 6 | 534 | 4163641 |