Gene Transcripts
- Gene Symbol
- Ilf2
- Entrez Gene ID
- 67781
- Gene Name
- interleukin enhancer binding factor 2
- Gene Aliases
- 6230405A16Rik, TEG-267, Tex261, Tex267
- Chromosome Location
- Chr. 3: 90476201 - 90488379 on Build GRCm38
- UniGene ID
- Mm.227258
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026374.3 |
NP_080650.1
|
5, 6 |
325 |
|
GenBank mRNA |
AK012364.1 |
- |
5, 6 |
312 |
|
AK013858.1 |
- |
5, 6 |
371 |
|
AK031892.1 |
- |
5, 6 |
380 |
|
AK044885.1 |
- |
5, 6 |
371 |
|
AK078003.1 |
- |
5, 6 |
312 |
|
AK135449.1 |
- |
5, 6 |
400 |
|
AK135539.1 |
- |
5, 6 |
312 |
|
AK157129.1 |
- |
5, 6 |
380 |
|
BC024718.1 |
- |
5, 6 |
325 |
5012365 |
View Full Product Details