Gene Transcripts

Gene Symbol
Pop5
Entrez Gene ID
117109
Gene Name
processing of precursor 5, ribonuclease P/MRP family (S. cerevisiae)
Gene Aliases
1500019J17Rik, 2700077E03Rik, AW049989, Rnasep3
Chromosome Location
Chr. 5: 115235851 - 115240972 on Build GRCm38
UniGene ID
Mm.24636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026398.4 NP_080674.1 4, 5 581
GenBank mRNA AK005290.1 - 3, 4 337
AK012421.1 - 3, 4 330
AK012523.1 - 339
AK013257.1 - 2, 3 318
BC022670.1 - 4, 5 605 4224973

View Full Product Details