Gene Transcripts

Gene Symbol
Spata6
Entrez Gene ID
67946
Gene Name
spermatogenesis associated 6
Gene Aliases
1700062C23Rik, AI790763, Hash, KRP, Mash
Chromosome Location
Chr. 4: 111719941 - 111829137 on Build GRCm38
UniGene ID
Mm.275526
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026470.3 NP_080746.3 5 591
GenBank mRNA AK051574.1 - 5 592
AK053170.1 - 5 565
AK132771.1 - 5 565
AK153098.1 - 5 468

View Full Product Details