Gene Transcripts
- Gene Symbol
- Rps13
- Entrez Gene ID
- 68052
- Gene Name
- ribosomal protein S13
- Gene Aliases
- 2700063M04Rik
- Chromosome Location
- Chr. 7: 116331507 - 116334190 on Build GRCm38
- UniGene ID
- Mm.14798
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026533.3 |
NP_080809.1
|
4, 5 |
412 |
|
GenBank mRNA |
AK012479.1 |
- |
4, 5 |
357 |
|
AK018703.1 |
- |
4, 5 |
356 |
|
AK076060.1 |
- |
4, 5 |
356 |
|
AK088237.1 |
- |
3, 4 |
989 |
|
AK151043.1 |
- |
4, 5 |
357 |
|
AK151671.1 |
- |
4, 5 |
358 |
|
AK166691.1 |
- |
4, 5 |
357 |
|
AK177445.1 |
- |
|
355 |
|
AV498110.1 |
- |
3, 4 |
319 |
|
BC011192.1 |
- |
4, 5 |
355 |
4016679 |
BC090397.1 |
- |
4, 5 |
339 |
5150054 |
BC106137.1 |
- |
4, 5 |
340 |
4990874 |
BC125602.1 |
- |
4, 5 |
356 |
40130117 |
BC125604.1 |
- |
4, 5 |
356 |
40130119 |
View Full Product Details