Gene Transcripts

Gene Symbol
Sult1c2
Entrez Gene ID
69083
Gene Name
sulfotransferase family, cytosolic, 1C, member 2
Gene Aliases
1810008N17Rik, ST1C1, ST1C2, Sult1c1
Chromosome Location
Chr. 17: 53829637 - 53845958 on Build GRCm38
UniGene ID
Mm.19320
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026935.4 NP_081211.3 2 306
GenBank mRNA AK007384.1 - 2 166
AK144336.1 - 2 246
AY005469.1 - 2 306
BC022665.1 - 2 299 4223678

View Full Product Details