Gene Transcripts

Gene Symbol
LAMA3
Entrez Gene ID
3909
Gene Name
laminin subunit alpha 3
Gene Aliases
BM600, E170, LAMNA, LOCS
Chromosome Location
Chr. 18: 23689443 - 23955066 on Build GRCh38
UniGene ID
Hs.436367
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000227.4 NP_000218.3 2 297
NM_001127717.2 NP_001121189.2 39 5340
NM_001127718.2 NP_001121190.2 2 297
NM_198129.2 NP_937762.2 39 5340
XM_011525978.2 XP_011524280.1 39 5367
XM_011525979.2 XP_011524281.1 39 5358
XM_011525980.2 XP_011524282.1 39 5349
XM_011525981.2 XP_011524283.1 38 5235
XM_011525982.2 XP_011524284.1 39 5367
XM_017025743.1 XP_016881232.1 22 3051
XM_017025744.1 XP_016881233.1 12 1732
GenBank mRNA AB107369.1 - 39 5099
AY327114.1 - 2 272
AY327115.1 - 39 5099
AY327116.1 - 39 5099
DA862923.1 - 2 297
L34155.1 - 2 239
X84392.1 - 2 274
X84900.1 - 4 518
X85107.1 - 2 290
X85108.1 - 4 518

View Full Product Details