Gene Transcripts

Gene Symbol
Zdhhc24
Entrez Gene ID
70605
Gene Name
zinc finger, DHHC domain containing 24
Gene Aliases
5730496N17Rik, Leng4
Chromosome Location
Chr. 19: 4878668 - 4885397 on Build GRCm38
UniGene ID
Mm.36885
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168516.1 NP_001161988.1 3 684
NM_001168517.1 NP_001161989.1 3 731
NM_027476.3 NP_081752.2 3 977
GenBank mRNA AK017737.1 - 3 942
AK028164.1 - 3 940
AK043923.1 - 3 934
AK169337.1 - 3 936
AK170233.1 - 3 961
AK170706.1 - 3 961
AK171568.1 - 3 961
AK172151.1 - 3 731
BC056621.1 - 3 699
BC071194.1 - 3 918
CD354313.1 - 3 640

View Full Product Details