Gene Transcripts

Gene Symbol
Rpap3
Entrez Gene ID
71919
Gene Name
RNA polymerase II associated protein 3
Gene Aliases
2310042P20Rik, D15Ertd682e
Chromosome Location
Chr. 15: 97675097 - 97705822 on Build GRCm38
UniGene ID
Mm.12255
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028003.2 NP_082279.1 7, 8 824
GenBank mRNA AK009765.1 - 7, 8 825
AK169097.1 - 7, 8 777
BC004046.1 - 7, 8 827 3496407

View Full Product Details