Gene Transcripts

Gene Symbol
Zdhhc13
Entrez Gene ID
243983
Gene Name
zinc finger, DHHC domain containing 13
Gene Aliases
2410004E01Rik, C530010M18, HIP3RP, Hip14l, kojak, skc4
Chromosome Location
Chr. 7: 48789003 - 48827440 on Build GRCm38
UniGene ID
Mm.279116
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028031.3 NP_082307.1 9, 10 1123
GenBank mRNA AK010382.1 - 9, 10 1123
AK049640.1 - 8, 9 952
AK145855.1 - 9, 10 1043
AK146196.1 - 9, 10 1095
BC019493.1 - 9, 10 1042
BC046599.1 - 9, 10 1042 6311721

View Full Product Details