Gene Transcripts

Gene Symbol
PEX6
Entrez Gene ID
5190
Gene Name
peroxisomal biogenesis factor 6
Gene Aliases
HMLR2, PAF-2, PAF2, PBD4A, PDB4B, PXAAA1
Chromosome Location
Chr. 6: 42963873 - 42979243 on Build GRCh38
UniGene ID
Hs.656425
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000287.3 NP_000278.3 9, 10 2047
NM_001316313.1 NP_001303242.1 9, 10 1783
NR_133009.1 9, 10 2047
XM_011514661.1 XP_011512963.1 8, 9 1963
GenBank mRNA AB051076.1 - 9, 10 1954
AB051077.1 - 9, 10 1690
AB051078.1 - 9, 10 1954
AB208906.1 - 9, 10 2007
AK314237.1 - 9, 10 1983
BC033487.1 - 9, 10 1962
BC048331.1 - 9, 10 1970 5189685
D83703.1 - 9, 10 2024

View Full Product Details