Gene Transcripts

Gene Symbol
Hspa12b
Entrez Gene ID
72630
Gene Name
heat shock protein 12B
Gene Aliases
2700081N06Rik
Chromosome Location
Chr. 2: 131127412 - 131145985 on Build GRCm38
UniGene ID
Mm.277465
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028306.3 NP_082582.1 5 465
GenBank mRNA AK012548.1 - 5 438
AY196790.1 - 4 345
BC011103.1 - 5 465 3990088

View Full Product Details