Gene Transcripts

Gene Symbol
Msl1
Entrez Gene ID
74026
Gene Name
male-specific lethal 1 homolog (Drosophila)
Gene Aliases
2810017F12Rik, 4121402D02Rik, 4930463F05Rik, AA682082, Msl-1
Chromosome Location
Chr. 11: 98795468 - 98807862 on Build GRCm38
UniGene ID
Mm.258352
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028722.2 NP_082998.2 3, 4 1452
GenBank mRNA AK014463.1 - 3, 4 1705
BC058629.1 - 3, 4 1452 6402215

View Full Product Details