Gene Transcripts

Gene Symbol
Syvn1
Entrez Gene ID
74126
Gene Name
synovial apoptosis inhibitor 1, synoviolin
Gene Aliases
1200010C09Rik, AW211966, C85322, D530017H19Rik, Hrd1
Chromosome Location
Chr. 19: 6045030 - 6053718 on Build GRCm38
UniGene ID
Mm.149870
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164709.1 NP_001158181.1 3 358
NM_028769.5 NP_083045.4 3 725
GenBank mRNA AK004688.2 - 3 212
AK122558.1 - 3 240
AK143720.1 - 3 237
AK172097.1 - 3 237
BC042199.1 - 3 220 4953165
BC046829.1 - 3 358 6811574
BC057917.1 - 3 639 5136398
BC080722.1 - 3 243 5323695

View Full Product Details