Gene Transcripts

Gene Symbol
Crct1
Entrez Gene ID
74175
Gene Name
cysteine-rich C-terminal 1
Gene Aliases
2300002G24Rik, AA589624, Nice-1
Chromosome Location
Chr. 3: 93014205 - 93015686 on Build GRCm38
UniGene ID
Mm.35806
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028798.3 NP_083074.2 2 645
GenBank mRNA AK009010.1 - 2 646
BC060280.1 - 2 624 30290660
CB202103.1 - 625

View Full Product Details