Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029561.4 | NP_083837.2 | 7, 8 | 1027 | |
GenBank mRNA | AK002978.1 | - | 5, 6 | 546 | |
AK031073.1 | - | 7, 8 | 842 | ||
AK049678.1 | - | 7, 8 | 897 | ||
AK078939.1 | - | 7, 8 | 937 | ||
AK132235.1 | - | 7, 8 | 956 | ||
AK150832.1 | - | 7, 8 | 841 | ||
BC071237.1 | - | 7, 8 | 946 | 5003738 | |
BC117075.1 | - | 7, 8 | 802 | 40126394 |