Gene Transcripts

Gene Symbol
Asb5
Entrez Gene ID
76294
Gene Name
ankyrin repeat and SOCs box-containing 5
Gene Aliases
1110018D09Rik
Chromosome Location
Chr. 8: 54550331 - 54587842 on Build GRCm38
UniGene ID
Mm.33171
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029569.3 NP_083845.1 7 1187
GenBank mRNA AF398966.1 - 7 1098
AK003770.1 - 7 1188
BC108405.1 - 7 1102 30288598
BC120851.1 - 7 1153 40129774
BC132007.1 - 7 1153 40130284

View Full Product Details