Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029891.2 | NP_084167.2 | 3 | 1467 | |
GenBank mRNA | AK042690.1 | - | 3 | 1469 | |
AK079123.1 | - | 1 | 248 | ||
BC095972.1 | - | 1 | 226 | ||
BC138828.1 | - | 3 | 1435 | 8861850 | |
BC138829.1 | - | 3 | 1435 | 8861851 |