Gene Transcripts

Gene Symbol
Gpr84
Entrez Gene ID
80910
Gene Name
G protein-coupled receptor 84
Gene Aliases
EX33
Chromosome Location
Chr. 15: 103308233 - 103311242 on Build GRCm38
UniGene ID
Mm.160369
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030720.1 NP_109645.1 2 1464
GenBank mRNA AF272948.1 - 2 1464
BC023249.1 - 2 1473 3995193

View Full Product Details