Gene Transcripts

Gene Symbol
Gtf2a1
Entrez Gene ID
83602
Gene Name
general transcription factor II A, 1
Gene Aliases
6330549H03Rik, AA536742, AA959775, AW060250, TfIIAa/b, Tfiia1
Chromosome Location
Chr. 12: 91555262 - 91590487 on Build GRCm38
UniGene ID
Mm.275728
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031391.2 NP_113568.2 7 787
NM_175335.3 NP_780544.1 7 774
GenBank mRNA AF250834.1 - 7 717
AK048711.1 - 7 775
AK139996.1 - 7 787
BC079909.1 - 7 720 30658355
BC117829.1 - 7 744 40092917
BC117830.1 - 7 744 40092921

View Full Product Details