Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031391.2 | NP_113568.2 | 7 | 984 | |
NM_175335.3 | NP_780544.1 | 7 | 971 | ||
GenBank mRNA | AF250834.1 | - | 7 | 914 | |
AK048711.1 | - | 7 | 972 | ||
BC079909.1 | - | 7 | 917 | 30658355 | |
BC117829.1 | - | 7 | 941 | 40092917 | |
BC117830.1 | - | 7 | 941 | 40092921 |