Gene Transcripts

Gene Symbol
Kars
Entrez Gene ID
85305
Gene Name
lysyl-tRNA synthetase
Gene Aliases
AA589550, AL024334, AL033315, AL033367, D8Ertd698e, D8Wsu108e, LysRS, mKIAA0070
Chromosome Location
Chr. 8: 111993439 - 112011354 on Build GRCm38
UniGene ID
Mm.196544
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130868.2 NP_001124340.1 3 379
NM_001286384.1 NP_001273313.1 4 437
NM_053092.3 NP_444322.1 2 199
GenBank mRNA AK090203.1 - 2 124
AK153236.1 - 2 123
AK167693.1 - 2 124
BC027356.1 - 4 349 4950333
BC035324.1 - 3 291 2811286
BC036289.1 - 2 168 3157706
BY792549.1 - 2 150

View Full Product Details