Gene Transcripts

Gene Symbol
Srd5a2
Entrez Gene ID
94224
Gene Name
steroid 5 alpha-reductase 2
Gene Aliases
5ART2, S5AR 2
Chromosome Location
Chr. 17: 74017706 - 74047916 on Build GRCm38
UniGene ID
Mm.38933
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053188.2 NP_444418.1 2 411
GenBank mRNA AB049456.1 - 2 411
AK138946.1 - 2 352
BC125510.1 - 2 395 40130025

View Full Product Details