Gene Transcripts

Gene Symbol
Crip3
Entrez Gene ID
114570
Gene Name
cysteine-rich protein 3
Gene Aliases
TLP, TLP-A, TLP-B, Tlmp
Chromosome Location
Chr. 17: 46428195 - 46431780 on Build GRCm38
UniGene ID
Mm.25168
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053250.2 NP_444480.1 8 744
NM_181664.2 NP_858050.1 8 685
GenBank mRNA AF367970.1 - 8 685
AF367971.1 - 8 744
AK080272.1 - 8 744
BC116360.1 - 8 685 40099973
BC116361.1 - 3 867

View Full Product Details