Gene Transcripts

Gene Symbol
Rfxap
Entrez Gene ID
170767
Gene Name
regulatory factor X-associated protein
Gene Aliases
5730495K23Rik
Chromosome Location
Chr. 3: 54803115 - 54807791 on Build GRCm38
UniGene ID
Mm.185735
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133231.2 NP_573494.1 1, 2 601
GenBank mRNA AF335512.1 - 1, 2 581
AK077640.1 - 1, 2 599
AK087489.1 - 1, 2 602
AK144633.1 - 1, 2 588
BC019935.1 - 1, 2 564 5041039
BC047123.1 - 1, 2 564

View Full Product Details