Gene Transcripts

Gene Symbol
Bco2
Entrez Gene ID
170752
Gene Name
beta-carotene oxygenase 2
Gene Aliases
B-diox-II, Bcdo2, Bcmo2, CMO2
Chromosome Location
Chr. 9: 50533087 - 50555220 on Build GRCm38
UniGene ID
Mm.390149
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133217.3 NP_573480.1 4 583
GenBank mRNA AJ290392.1 - 3 426
BC107006.2 - 3 426 40046913
BC107007.2 - 3 426 40046914

View Full Product Details